UV PCR Chambers
Isolation chamber provides protection against Beta radiation
  • Airtight chamber reduces the chance of airborne contamination
  • UV germicidal lamps decontaminate interior chamber
This chamber improves the accuracy of PCR and general tissue culture procedures by greatly reducing the chance of airborne contamination during DNA sequencing. Airtight chamber features a built-in UV system rated at 254 nm to decontaminate interior surfaces. Includes fluorescent and UV germicidal lamps.
Removable doors save valuable laboratory countertop space. If a corner door is opened, electrical sensors turn off the UV system. Overall chamber dimensions are 24''W x 28''H x 18''D.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Interior Depth (in) Interior Width (in) Power (VAC)
Availability Pricing
Plas-Labs 825-UVC UV PCR Chamber, 110 VAC 3475075 Mfr # 825-UVC/110 (PCR)
COMPARE
18 24 110
Call For Price
Plas-Labs 825-UVC UV PCR Chamber, 230 VAC 3475077 Mfr # 825-UVC/EXP
COMPARE
18 24 220