UVP PhotoDoc-It™ Imaging Systems
Compact system quickly prints and shoots brilliant color gel images with a high resolution camera
  • Instantly print color images on the photo printer included with the system
  • Save images to the memory card or connect to a computer via USB (not included)
  • Select systems include UV transilluminator
  • Ethidium bromide filter for fluorescent-stained gels
System features a high-resolution, 16 megapixel digital color camera for capturing brilliant color images. Lightweight, durable hood enclosure provides a compact darkroom environment. The ethidium bromide emission filter, positioned inside the hood, is easily interchangeable for other filter types.
Models 97700-14 through -28 include a benchtop UV transilluminator to illuminate stained gels with the UV fluorescence. Color printer generates instant 4" x 6" (10.2 x 15.2 cm) color prints. Three-digit resettable combination lock secures the camera to the hood.
598,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Filter Wavelength (nm) Power (VAC) Description
Availability Pricing
PhotoDoc-It UVP 50 Imaging System; 230 VAC 9770012 Mfr # 97-0274-02
COMPARE
230 UVP 50 Imaging System; 230 VAC
₹598,000.00INR / Each
PhotoDoc-It UVP Imaging System, 302/365nm 20x20cm Transilluminator; 230V 9770020 Mfr # 97-0274-08
COMPARE
302/365 230 UVP Imaging System, 302/365nm 20x20cm Transilluminator; 230V