Value Plastics Female Luer to 500 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The taper of the female luer is designed to match the taper of the male luer, creating a secure and leak-free connection when the two parts are joined together. The 500 series barb is composed of blended radii giving it a distinct look. Each barb is designed to accommodate a range of tube sizes and tube types. Consideration should be given to flow restriction, pressure capability, and the force required for assembly of the tube. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Options include nylon, polycarbonate, or polypropylene resin.
Common features of nylon include a high mechanical strength, great balance of stiffness and toughness, good performance at high temperatures, good electrical and flammability properties, and resistance against abrasion or chemicals. It can be sterilized using gamma radiation, Ethylene oxide (EtO), or autoclave.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
Polycarbonate is transparent and features high impact, chemical, UV, and electrical resistance. Sterilization is only recommended via ethylene oxide (EtO). Gamma radiation up to 10 Mrads is possible with minor loss of physical properties – it will discolor to a yellow-green hue. Autoclave is not recommended and may crack the fitting.
1,700.00 - ₹2,700.00INR / Pkg of 25
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size-Port 2 (in) Connection Type-Port 1 Material
Availability Pricing
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Female Luer Thread, Smooth Bore x 500 Series Barb 1/8" ID; 25/Pk 5011996 Mfr # FTLLSB013-6005
₹2,200.00INR / Pkg of 25
COMPARE
1/8 Female Luer Polypropylene
Loading
₹2,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Female Luer Thread x 500 Series Barb 1/8" ID; 25/Pk 5012036 Mfr # FTLL013-1
₹1,700.00INR / Pkg of 25
COMPARE
1/8 Female Luer Nylon
Loading
₹1,700.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Female Luer Thread x 500 Series Barb 1/16" ID; 25/Pk 5012042 Mfr # FTLL004-6005
₹2,200.00INR / Pkg of 25
COMPARE
1/16 Female Luer Polypropylene
Loading
₹2,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread x 500 Series Barb 1/4" ID; 25/Pk 5012043 Mfr # FTLL055-9
₹2,400.00INR / Pkg of 25
COMPARE
1/4 Female Luer Polycarbonate
Loading
₹2,400.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread x 500 Series Barb 1/16" ID; 25/Pk 5012048 Mfr # FTLL004-9
₹2,100.00INR / Pkg of 25
COMPARE
1/16 Female Luer Polycarbonate
Loading
₹2,100.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Female Luer Thread x 500 Series Barb 1/8" ID; 25/Pk 5012054 Mfr # FTLL013-6005
₹2,200.00INR / Pkg of 25
COMPARE
1/8 Female Luer Polypropylene
Loading
₹2,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Female Luer Thread x 500 Series Barb 1/16" ID; 25/Pk 5012061 Mfr # FTLL004-1
₹1,700.00INR / Pkg of 25
COMPARE
1/16 Female Luer Nylon
Loading
₹1,700.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Female Luer Thread x 500 Series Barb 1/4" ID; 25/Pk 5012062 Mfr # FTLL055-6005
₹2,700.00INR / Pkg of 25
COMPARE
1/4 Female Luer Polypropylene
Loading
₹2,700.00INR / Pkg of 25
Showing 1- 8 of 8