Value Plastics Sanitary to 600 Series Hose Barb Fittings
Sanitary fittings provide a secure connection where preventing contamination is crucial
  • Reduce the risk of bacterial growth with smooth surfaces and minimal crevices
  • Easily transition from sanitary to hose barb connection
  • Maintain a low profile and ease of assembly with 600 series barb fittings
These Value Plastics Sanitary to 600 Series Barb fittings are designed to easily connect sanitary ports to flexible tubing. Sanitary fittings ensure a reliable connection while facilitating easy cleaning and preventing the buildup of bacteria or other contaminants. The 600 Series barbs are designed for use with flexible tubing commonly used in food and beverage, pharmaceutical, and biotechnology applications. Tubing is expanded from 18% to 34% above its nominal ID to maintain a low profile and ease of assembly, and the straight barb stem facilitates use of recommended cable tie retainers or hose clamps. These adapters are available in various materials to accommodate various applications and compliance requirements.
147,000.00 - ₹248,000.00INR / Pkg of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Sanitary Clamp Size (Port 1) (in)
Availability Pricing
Value Plastics Fitting, Sanitary to 600 Series Barb Adapter, Straight, Polypropylene, Mini 3/4" Sanitary x 3/4" ID; 100/Pk 5011864 Mfr # SFMN6100-6005
COMPARE
3/4 Polypropylene 0.75
₹175,000.00INR / Pkg of 100
Value Plastics Fitting, Sanitary to 600 Series Barb Adapter, Straight, Polypropylene, Mini 3/4" Sanitary x 3/8" ID; 100/Pk 5011865 Mfr # SFMN670-6005
COMPARE
3/8 Polypropylene 0.75
₹147,000.00INR / Pkg of 100
Value Plastics Fitting, Sanitary to 600 Series Barb Adapter, Straight, Polypropylene, Mini 3/4" Sanitary x 1/2" ID; 100/Pk 5011866 Mfr # SFMN680-6005
COMPARE
1/2 Polypropylene 0.75
₹150,000.00INR / Pkg of 100
Value Plastics Fitting, Sanitary to 600 Series Barb Adapter, Straight, Polypropylene, 1-1/2" Sanitary x 3/4" ID; 100/Pk 5011868 Mfr # SFMX6100-6005
COMPARE
3/4 Polypropylene 1.5
₹203,000.00INR / Pkg of 100
Value Plastics Fitting, Sanitary to 600 Series Barb Adapter, Straight, PVDF, 1-1/2" Sanitary x 600 Series Barb 1"ID; 100/Pk 5011869 Mfr # SFMX6110-J2A
COMPARE
1 PVDF 1.5
₹248,000.00INR / Pkg of 100
Value Plastics Fitting, Sanitary to 600 Series Barb Adapter, Straight, Polypropylene, 1-1/2" Sanitary x 1/4" ID; 100/Pk 5011870 Mfr # SFMX655-6005
COMPARE
1/4 Polypropylene 1.5
₹150,000.00INR / Pkg of 100
Value Plastics Fitting, Sanitary to 600 Series Barb Adapter, Straight, Polypropylene, 1-1/2" Sanitary x 3/8" ID; 100/Pk 5011871 Mfr # SFMX670-6005
COMPARE
3/8 Polypropylene 1.5
₹153,000.00INR / Pkg of 100
Showing 1- 7 of 7