Vivaspin 20 Centrifugal Concentrators with PES MWCO Membranes
Broad range of process alternatives for the filtration and concentration of biological samples
  • Disposable ultrafiltration devices concentrate and purify biological samples to concentrations over 100x in a single spin!
  • Concentrators feature twin vertical membranes—increased surface area provides faster filtration
  • Polyethersulfone membrane is excellent for most solutions when recovery is most important
Vivaspin 20 handles up to 20 ml in swing bucket centrifuges and 14 ml in fixed angle rotors accepting 50 ml centrifuge tubes.
39,900.00 - ₹149,000.00INR / Pkg of 12
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Max Molecular Weight Cut Off (Mwco) (Dalton) Description
Availability Pricing
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 5000 Da; 12/Pk 3622470 Mfr # VS2011
COMPARE
5000 Centrifugal Concentrators, PES Membrane 5000 Da; 12/Pk
₹39,900.00INR / Pkg of 12
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 5000 Da; 48/Pk 3622471 Mfr # VS2012
COMPARE
5000 Centrifugal Concentrators, PES Membrane 5000 Da; 48/Pk
₹145,000.00INR / Pkg of 48
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 10,000 Da; 12/Pk 3622472 Mfr # VS2001
COMPARE
10000 Centrifugal Concentrators, PES Membrane 10,000 Da; 12/Pk
₹39,900.00INR / Pkg of 12
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 10,000 Da; 48/Pk 3622473 Mfr # VS2002
COMPARE
10000 Centrifugal Concentrators, PES Membrane 10,000 Da; 48/Pk
₹145,000.00INR / Pkg of 48
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 30,000 Da; 12/Pk 3622474 Mfr # VS2021
COMPARE
30000 Centrifugal Concentrators, PES Membrane 30,000 Da; 12/Pk
₹39,900.00INR / Pkg of 12
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 30,000 Da; 48/Pk 3622475 Mfr # VS2022
COMPARE
30000 Centrifugal Concentrators, PES Membrane 30,000 Da; 48/Pk
₹145,000.00INR / Pkg of 48
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 50,000 Da; 12/Pk 3622476 Mfr # VS2031
COMPARE
50000 Centrifugal Concentrators, PES Membrane 50,000 Da; 12/Pk
₹39,900.00INR / Pkg of 12
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 50,000 Da; 48/Pk 3622477 Mfr # VS2032
COMPARE
50000 Centrifugal Concentrators, PES Membrane 50,000 Da; 48/Pk
₹145,000.00INR / Pkg of 48
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 100,000 Da; 12/Pk 3622478 Mfr # VS2041
COMPARE
100000 Centrifugal Concentrators, PES Membrane 100,000 Da; 12/Pk
₹41,100.00INR / Pkg of 12
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 100,000 Da; 48/Pk 3622479 Mfr # VS2042
COMPARE
100000 Centrifugal Concentrators, PES Membrane 100,000 Da; 48/Pk
₹149,000.00INR / Pkg of 48
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 0. um; 48/Pk 3622481 Mfr # VS2072
COMPARE
Centrifugal Concentrators, PES Membrane 0. um; 48/Pk
₹144,000.00INR / Pkg of 48
Showing 1- 11 of 11