Watson-Marlow Bioprene® TPE Pump Tubing
Provides accurate and reliable fluid transfer—ideal for critical processes
  • Produced from all synthetic components
  • Offers exceptionally long service life
  • Manufactured in an ISO 114644-1 Class 7 Cleanroom
  • Meets USP Class VI and FDA CFR 177.2600
  • Made from NSF 51 food compliant raw material
  • Opaque tubing offers protection from visible and UV light
  • Sterilize by autoclave or gamma irradiation
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Use the USP Class VI and FDA-compliant Bioprene TPE tubing in various biopharmaceutical and food applications. Produced from synthetic components, the tubing does not contain any phthalates or latex. This durable tubing offers an exceptionally long service life in peristaltic pumps, minimizing downtime from tubing changes. Tubing is manufactured and double-packed in heat-sealed polyethylene bags in an ISO Class 7 cleanroom to ensure that the integrity is not compromised.
14,100.00 - ₹30,400.00INR / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (m) Tubing ID (mm) Wall Thickness (mm)
Availability Pricing
Watson-Marlow Bioprene® TPE Pump Tubing, 0.5 mm Bore x 1.6 mm Wall; 5 m 5013027 Mfr # 933.0005.J16
COMPARE
5 0.5 1.6
₹14,100.00INR / Pkg of 1
Watson-Marlow Bioprene® TPE Pump Tubing, 0.8 mm Bore x 1.6 mm Wall; 5 m 5013028 Mfr # 933.0008.J16
COMPARE
5 0.8 1.6
₹16,100.00INR / Pkg of 1
Watson-Marlow Bioprene® TPE Pump Tubing, 1.6 mm Bore x 1.6 mm Wall; 5 m 5013029 Mfr # 933.0016.J16
COMPARE
5 1.6 1.6
₹20,300.00INR / Pkg of 1
Watson-Marlow Bioprene® TPE Pump Tubing, 2.4 mm Bore x 1.6 mm Wall; 5 m 5013030 Mfr # 933.0024.J16
COMPARE
5 2.4 1.6
₹20,500.00INR / Pkg of 1
Watson-Marlow Bioprene® TPE Pump Tubing, 3.2 mm Bore x 1.6 mm Wall; 5 m 5013031 Mfr # 933.0032.J16
COMPARE
5 3.2 1.6
₹21,500.00INR / Pkg of 1
Watson-Marlow Bioprene® TPE Pump Tubing, 4.8 mm Bore x 1.6 mm Wall; 5 m 5013032 Mfr # 933.0048.J16
COMPARE
5 4.8 1.6
₹23,000.00INR / Pkg of 1
Watson-Marlow Bioprene® TPE Pump Tubing, 6.4 mm Bore x 1.6 mm Wall; 5 m 5013033 Mfr # 933.0064.J16
COMPARE
5 6.4 1.6
₹26,700.00INR / Pkg of 1
Watson-Marlow Bioprene® TPE Pump Tubing, 8.0 mm Bore x 1.6 mm Wall; 5 m 5013034 Mfr # 933.0080.J16
COMPARE
5 8 1.6
₹27,600.00INR / Pkg of 1
Watson-Marlow Bioprene® TPE Pump Tubing, 4.0 mm Bore x 1.6 mm Wall; 10 m 5014035 Mfr # 933.0040.016
COMPARE
10 4 1.6
₹30,400.00INR / Pkg of 1
Showing 1- 9 of 9