Weighted Tubing Closures for Spectra/Por RC Membranes
The most convenient method for sealing dialysis tubing without the risk of leaking
  • Easy to open and reseal
Weighted closures feature a fluorocarbon-coated stainless steel bar. The weighted closure acts like an anchor to maintain a vertical orientation of the sample.
33,000.00 - ₹37,700.00INR / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Color Tubing Sealing Width (mm)
Availability Pricing
Spectra Por 132742 RC Dialysis Membrane Tubing Closure-Weighted, 12MM White 10/pk 0289917 Mfr # 132742
COMPARE
White 12
₹33,000.00INR / Pkg of 10
Spectra Por 132746 RC Dialysis Membrane Tubing Closure-Weighted, 75MM White 10/pk 0289927 Mfr # 132746
COMPARE
White 75
₹37,700.00INR / Pkg of 10
Spectra Por 132744 Weighted Tubing Closures; max gripping width, 35mm; 10/box 0289954 Mfr # 132744
COMPARE
White 35
₹35,300.00INR / Box of 10
Spectra Por 132745 Weighted Tubing Closures; max gripping width, 55mm; 10/box 0289956 Mfr # 132745
COMPARE
White 55
₹36,100.00INR / Box of 10