WIKA A-10 Pressure Transmitter
  • Pressure ranges up to 10,000 psi
  • Non-linearity is < + 0.5% BFSL
  • Signal output is 4 to 20 mA
  • Electrical connection- DIN 43 650 plug connector
The WIKA A-10 pressure transmitter is precision engineered and manufactured to fit many industrial and OEM pressure measurement applications. The rugged design provides resistance to vibration, shock, wide temperature variations, RFI and other extreme environmental conditions that are typical of industrial and OEM applications. Performance and reliability is enhanced by the all stainless steel welded measuring cell that eliminates the need for soft sealing materials that may deteriorate over time. The state-of-the-art manufacturing and assembly process increases the long term reliability of the A-10.
Primary applications include process control and automation, hydraulics, pneumatics, and machine controls.
36,000.00 - ₹45,400.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Output Process Connection
Availability Pricing
Wika A-10 Pressure Transmitter, 0 To 15 PSIA; 1/4" NPT(M) Connection 6806201 Mfr # 50426354
COMPARE
29.7 4 to 20 mA 1/4" NPT(M)
₹45,400.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 100 PSIA; 1/4" NPT(M) Connection 6806203 Mfr # 50426389
COMPARE
114.7 4 to 20 mA 1/4" NPT(M)
₹45,400.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 15 PSI; 1/4" NPT(M) Connection 6806205 Mfr # 50426397
COMPARE
15 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika 54626401 Pressure Transmitter, 0 To 25 PSI; 1/4" NPT(M) Connection 6806207 Mfr # 50426401
COMPARE
25 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 50 PSI; 1/4" NPT(M) Connection 6806209 Mfr # 50426427
COMPARE
50 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 100 PSI; 1/4" NPT(M) Connection 6806211 Mfr # 50372475
COMPARE
100 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 200 PSI; 1/4" NPT(M) Connection 6806213 Mfr # 50398083
COMPARE
200 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 300 PSI; 1/4" NPT(M) Connection 6806215 Mfr # 50426460
COMPARE
300 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 500 PSI; 1/4" NPT(M) Connection 6806217 Mfr # 50426478
COMPARE
500 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 1,000 PSI; 1/4" NPT(M) Connection 6806219 Mfr # 50426486
COMPARE
1000 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 3,000 PSI; 1/4" NPT(M) Connection 6806225 Mfr # 50426516
COMPARE
3000 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 5,000 PSI; 1/4" NPT(M) Connection 6806227 Mfr # 50372483
COMPARE
5000 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Wika A-10 Pressure Transmitter, 0 To 10,000 PSI; 1/4" NPT(M) Connection 6806229 Mfr # 50426532
COMPARE
10000 4 to 20 mA 1/4" NPT(M)
₹36,000.00INR / Each
Showing 1- 13 of 13