Wika Industrial Pressure Gauges, 4-1/2" Dial
Designed for large-volume use—such as in the heating/air conditioning field
  • Use with any liquid or gas not corrosive to copper alloys
  • Accuracy is ±1% (ASME Grade 1A)
Gauges feature copper alloy tube, brass socket, 304 stainless steel case, and plastic lens. Bottom connection is 1/4" NPT(M).
7,100.00 - ₹7,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Dial Size (in) Max Pressure (PSI)
Availability Pricing
Wika 111.25 4.5 0 to 60 psi Industrial Gauge, 4 1/2" Dial 6804714 Mfr # 4277776
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 60
₹7,100.00INR / Each
Wika 111.25 4.5 0 to 100 psi Industrial Gauge, 4 1/2" Dial 6804716 Mfr # 4277784
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 100
₹7,500.00INR / Each