WIKA S-20 Rugged Harsh-Environment Pressure Transmitters
An exceptional choice for many demanding process applications
  • 4 to 20 mA output
  • Accuracy of ±0.5% full-scale
Designed for use in demanding environments, the WIKA S-20 transmitters feature high accuracy and a rugged design. These units are very reliable in harsh environments, including extreme shock, high vibration, and aggressive media, as well as general industrial applications. Versatile 4-pin DIN electrical connections provide easy wiring. Models with a 1/2" NPT(M) process connection have a standard DIN connection; models with a 1/4" NPT(M) process connection have a mini-DIN electrical connection.
84,600.00 - ₹152,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Min Pressure (PSI) Process Connection
Availability Pricing
Wika S-20 SS IP65 Vacuum Transmitter, 30-0"Hg, 1/2" NPT 6808300 Mfr # 52375595
COMPARE
14.735 0 1/2" NPT(M)
₹152,000.00INR / Each
Wika S-20 SS IP65 Pressure Transmitter, 0-60 psi, 1/2" NPT 6808308 Mfr # 52375293
COMPARE
60 0 1/2" NPT(M)
₹128,000.00INR / Each
Wika S-20 SS IP65 Pressure Transmitter, 0-500 psi, 1/2" NPT 6808318 Mfr # 52375366
COMPARE
500 0 1/2" NPT(M)
₹128,000.00INR / Each
Wika S-20 SS IP65 Vacuum Transmitter, 30-0"Hg, 1/4" NPT 6808328 Mfr # 52377199
COMPARE
14.735 0 1/4" NPT(M)
₹109,000.00INR / Each
Wika S-20 SS IP65 Pressure Transmitter, 0-10 psi, 1/4" NPT 6808332 Mfr # 52377563
COMPARE
10 0 1/4" NPT(M)
₹105,000.00INR / Each
Wika S-20 SS IP65 Pressure Transmitter, 0-200 psi, 1/4" NPT 6808342 Mfr # 52376591
COMPARE
200 0 1/4" NPT(M)
₹84,600.00INR / Each
Wika S-20 SS IP65 Pressure Transmitter, 0-300 psi, 1/4" NPT 6808344 Mfr # 52376613
COMPARE
300 0 1/4" NPT(M)
₹84,600.00INR / Each
Wika S-20 SS IP65 Pressure Transmitter, 0-500 psi, 1/4" NPT 6808346 Mfr # 52376648
COMPARE
500 0 1/4" NPT(M)
₹84,600.00INR / Each
Wika S-20 SS IP65 Pressure Transmitter, 0-10000 psis, 1/4" NPT 6808354 Mfr # 52376834
COMPARE
10000 0 1/4" NPT(M)
₹84,600.00INR / Each
Showing 1- 9 of 9