WTW ProfiLine Cond 3110 Handheld Conductivity Meters
A rugged new line of meters with a range of features, benefits, and price points to meet every application and budget!
  • Designed for users who desire a simple-to-use, yet dependable instrument
  • Rugged, waterproof housing is great for the field or the lab
Durable, I67-rated meters feature a 100% waterproof and easy-to-clean keypad. Silicone rubber keys provide tactile feedback for error-free operation. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timer ensures more reliable results.
Meters include: CD-ROM and four AA batteries.
Meter kits add: specified probe, calibration solutions, stand, beaker, and hard carrying case.
198,000.00 - ₹335,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
WTW 2CA100 ProfiLine Cond 3110 Handheld Conductivity Meter Only 1970642 Mfr # 2CA100
COMPARE
ProfiLine Cond 3110 Handheld Conductivity Meter Only
₹198,000.00INR / Each
WTW 2CA100 Cond 3110 meter kit and TetraCon probe, 1-m cable 1970643 Mfr # 2CA101
COMPARE
Cond 3110 meter kit and TetraCon probe, 1-m cable
₹335,000.00INR / Each