WTW ProfiLine Cond 3310 Handheld Conductivity Meters
A rugged line of conductivity meters with a range of features, benefits, and price points to meet every application and budget!
  • Innovative waterproof USB connection for ultra-flexible data output
  • Rugged, waterproof housing is great for the field or the lab
Durable, I67-rated meters feature a 100% waterproof and easy-to-clean keypad. Silicone rubber keys provide tactile feedback for error-free operation. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timer ensures more reliable results.
Meter provide extended data memory for logging up to 5000 GLP-compliant data sets. Time-controlled datalogging function offers adjustable data collection intervals—ideal for long-term field monitoring. A special “sleep” mode saves power and extends battery life when performing long-term monitoring. Nonvolatile memory saves all stored data even if battery power is lost. Data transfer to PC is made
simple via the unique waterproof mini-USB interface.
Meters include: CD-ROM and four AA batteries.
Meter kits add: specified probe, calibration solutions, stand, beaker, and hard carrying case.
217,000.00 - ₹346,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
WTW 2CA300 ProfiLine Cond 3310 Handheld Conductivity Meter Only 1970650 Mfr # 2CA300
COMPARE
ProfiLine Cond 3310 Handheld Conductivity Meter Only
₹217,000.00INR / Each
WTW 2CA301 Cond 3310 meter kit and TetraCon probe, 1-m cable 1970651 Mfr # 2CA301
COMPARE
Cond 3310 meter kit and TetraCon probe, 1-m cable
₹346,000.00INR / Each