Environmental Express Air-O-Cell® Air Sampling Cassettes
Specifically designed for fast collection of airborne aerosols and inorganic particulates
  • No need to prepare slides or worry about contamination
  • Special adhesive on collection media prevents collected sample from being washed off
  • Attain direct quantitative analysis of organic and inorganic particulate
Use these disposable, easy-to-use air sampling cassettes to collect a wide range of airborne particulates such as mold, spores, pollen, insect parts, fibers, skin cells, and more. Air-O-Cell cassettes provide a broader overview of potential contaminants than conventional sampling methods by collecting both viable and nonviable sample specimens.
Connect to an air sampling pump to draw particulate-laden sample through the tapered inlet towards a slide. Slide features special collection media with adhesive to prevent particulate from blurring or being washed off during the staining process. Once the sampling is complete, send the Air-O-Cell cassettes to a laboratory, where the slides are removed and analyzed preventing contamination worries.
7,100.00 - ₹28,300.00INR / Box of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Cassettes, Air-O-Cell®; 10/BX
₹7,100.00INR / Box of 10
COMPARE
Cassettes, Air-O-Cell®; 50/BX
₹28,300.00INR / Box of 50