Zefon Aluminum Cyclone Calibration Adapter
Help ensure flow rate accuracy
  • Fits on both 25- and 37-mm aluminum cyclones
Use a calibration adapter to ensure the flow rate accuracy. Place the adapter over the stem of a 25- or 37-mm cyclone then connect to a flow calibrator using a 1/4" ID tubing. This allows an actual measurement as close to the cyclone inlet as possible for the most accurate reading.
3,300.00 - ₹9,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
COMPARE
Aluminum 25- and 37-mm aluminum cyclones Zefon Calibration Adapter, Aluminum Cyclone
₹9,800.00INR / Each
COMPARE
Aluminum cyclone calibration adapter Zefon O-Ring, Calibration Adapter, Aluminum Cyclone; 5/PK
₹3,300.00INR / Pkg of 5