Zefon High-Volume Diaphragm Sampling Pump
Perfect pump for use with asbestos PCM and TEM cassettes, carpet sampling cassettes, and Air-O-Cell® IAQ sampling cassettes
  • Flow range of 3 to 14 L/min
  • 23 strokes/min
55,400.00 - ₹187,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Environmental Express Diaphragm Pump, High Volume; 115V 6006211
₹56,100.00INR / Each
COMPARE
Diaphragm Pump, High-Volume; 115 VAC
Loading
₹56,100.00INR / Each
Environmental Express Diaphragm Pump, High Volume; 220V 6006212
₹55,400.00INR / Each
COMPARE
Diaphragm Pump, High-Volume; 220 VAC
Loading
₹55,400.00INR / Each
Environmental Express Diaphragm Pump, High Volume, 2 Pump Kit; 120V 6006213
₹182,000.00INR / Each
COMPARE
Diaphragm Pump, High-Volume, 2 Pump Kit; 120 VAC
Loading
₹182,000.00INR / Each
Environmental Express Diaphragm Pump, High Volume, 2 Pump Kit; 220V 6006214
₹187,000.00INR / Each
COMPARE
Diaphragm Pump, High-Volume, 2 Pump Kit; 220 VAC
Loading
₹187,000.00INR / Each