Zefon Mixed Cellulose Ester (MCE) Filter Membranes
Hydrophilic membranes—excellent for microbiology, contamination analysis, and air sampling applications
  • Developed from a mixture of cellulose acetate and cellulose nitrate
  • Dissolves completely using standard digestion procedures
  • Max operating temperature of 131°F (55°C)
  • Protein binding: 150 µg/cm2
  • Bacterial endotoxins: 8.0 EU/mL
Use hydrophilic MCE filter membranes for both air monitoring and analytical applications. Membranes clear completely, possess low artifacts, and offer minimal interference in fiber counting. MCE membranes are suitable for use in most NIOSH analytical methods, including asbestos fibers monitoring.
5,400.00 - ₹17,000.00INR / Pkg of 100
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (µm) Description
Availability Pricing
COMPARE
25 0.8
₹7,300.00INR / Pkg of 100
COMPARE
25 1.2
₹8,900.00INR / Pkg of 100
COMPARE
25 0.45
₹6,800.00INR / Pkg of 100
COMPARE
37 0.45
₹9,600.00INR / Pkg of 100
COMPARE
37 0.22
₹15,100.00INR / Pkg of 100
COMPARE
47 0.22
₹17,000.00INR / Pkg of 100
COMPARE
25 0.45
₹6,600.00INR / Pkg of 100
COMPARE
37 0.45
₹10,100.00INR / Pkg of 100
COMPARE
47 0.45
₹15,000.00INR / Pkg of 100
COMPARE
47 5
₹13,400.00INR / Pkg of 100
COMPARE
25 5
₹7,200.00INR / Pkg of 100
COMPARE
37 5
₹10,400.00INR / Pkg of 100
COMPARE
25 0.8
₹5,400.00INR / Pkg of 100
COMPARE
37 0.8
₹7,800.00INR / Pkg of 100
COMPARE
47 0.8
₹14,800.00INR / Pkg of 100
Showing 1- 15 of 15