Zefon Preloaded Air Sampling Cassettes with Polycarbonate Track-Etch (PCTE) Filter Membrane
PCTE membranes have uniform pore sizes, high flow rates, and excellent chemical and thermal resistance
  • Fully assembled and preloaded with 37-mm diameter PCTE membrane
  • Leak-free design—eliminates sample loss
  • Clear styrene housing provides excellent visibility of sample
  • Conform to most NIOSH standards
Make air sampling fast and simple with preassembled, preloaded filter cassettes. Cassettes come preloaded with PCTE membranes that are transparent and nonstaining. They feature a smooth surface and well-controlled pore size and shape, making them ideal for asbestos testing and scanning electron microscopy applications. Select PCTE membranes with porous plastic pad when sampling chemicals that are not compatible with cellulose. Cassettes are preloaded in an ISO 9001:2015-certified facility ensuring high quality control standards.
17,400.00 - ₹36,800.00INR / Box of 50
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Pore Size (µm) Support Description
Availability Pricing
COMPARE
0.4 Cellulose Cassette, Preloaded, 37 mm, 2 Piece, Styrene, 0.4 µm, Polycarbonate, Cellulose Pad; 50/BX
₹17,400.00INR / Box of 50
COMPARE
0.8 Cellulose Cassette, Preloaded, 37 mm, 2 Piece, Styrene, 0.8 µm, Polycarbonate, Cellulose Pad; 50/BX
₹17,400.00INR / Box of 50
COMPARE
0.4 Cellulose Cassette, Preloaded, 37 mm, 3 Piece, Styrene, 0.4 µm, Polycarbonate, Cellulose Pad; 50/BX
₹22,500.00INR / Box of 50
COMPARE
0.8 Cellulose Cassette, Preloaded, 37 mm, 3 Piece, Styrene, 0.8 µm, Polycarbonate, Cellulose Pad; 50/BX
₹36,800.00INR / Box of 50
COMPARE
0.8 Porous plastic Cassette, Preloaded, 37 mm, 3 Piece, Styrene, 0.8 µm, Polycarbonate, Plastic Pad; 50/BX
₹27,800.00INR / Box of 50