Zefon Preloaded Air Sampling Cassettes with Solu-Sert™ Filter Capsules
Captures 100% of sample collected within the capsule
  • Fully assembled and preloaded with 0.8 µm Solu-Sert filter capsule
  • Leak-free design—eliminates sample loss
  • Conform to most NIOSH standards
  • Two-piece, clear styrene housing
Make air sampling fast and simple with preassembled, preloaded filter cassettes. Cassettes come preloaded with Solu-Sert™ capsule to eliminate the need for wiping or rinsing. Capsule consists of an MCE filter membrane sealed to a cellulose shell to capture 100% of collected sample, including wall deposits. It is suitable for collecting aluminum, arsenic, chromium, lead, and other airborne metal contaminants under NIOSH Method 7306. Cassettes are preloaded in an ISO 9001:2015-certified facility, ensuring high quality control standards.
Clear styrene housing lets you view the filter media. Model with band provides added leak protection as well as offer a place to write notes.
34,900.00INR / Box of 50
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (µm) Description
Availability Pricing
COMPARE
25 0.8 Cassette, Preloaded, 25 mm, 2 Piece, Styrene, 0.8 µm, Solu-Sert™; 50/BX
₹54,800.00INR / Box of 50
COMPARE
37 0.8 Cassette, Preloaded, 37 mm, 2 Piece, Styrene, 0.8 µm, Solu-Sert™; 50/BX
₹32,700.00INR / Box of 50
COMPARE
37 0.8 Cassette, Preloaded, 37 mm, 2 Piece, Styrene, 0.8 µm, Solu-Sert™, White Band; 50/BX
₹34,900.00INR / Box of 50