Zefon Preloaded PCM Air Sampling Cassettes
Meet NIOSH, OSHA, and AHERA standards for asbestos air monitoring
  • PCM cassettes come preloaded, ready for use
  • Offers fast turnaround and low cost
  • Each cassette marked with a unique serial number
  • 2" static conductive extension cowl
Test for asbestos presence in the air with PCM cassettes. Each 25-mm, three-piece housing is injection molded in-house from high-quality polypropylene and is individually assembled and preloaded under stringent quality control standards, ensuring you get accurate results. Once assembled, each cassette undergoes an inspection for the correct filter/pad combination before being shipped out. Random lot sample testing is performed daily by an independent laboratory to ensure low background fiber counts. Laboratory results are available upon request.
8,100.00 - ₹36,500.00INR / Box of 50
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Pore Size (µm) Description
Availability Pricing
COMPARE
Polycarbonate 0.4 Cassette, 25 mm, 3 Piece, 0.4 µm, Polycarb; 50/Bx
₹36,500.00INR / Box of 50
COMPARE
Mixed cellulose ester 0.8 Cassette, 25 mm, 3 Piece, 0.8 µm, MCE; 50/BX
₹8,100.00INR / Box of 50
COMPARE
Mixed cellulose ester 0.8 Cassette, 25 mm, 3 Piece, 0.8 µm, MCE with Grids; 50/BX
₹12,500.00INR / Box of 50
COMPARE
Polycarbonate 0.8 Cassette, 25 mm, 3 Piece, 0.8 µm, Polycarbonate; 50/BX
₹14,500.00INR / Box of 50
COMPARE
Mixed cellulose ester 1.2 Cassette, 25 mm, 3 Piece, 1.2 µm, MCE with Grids; 50/BX
₹20,300.00INR / Box of 50