Zefon Silver Membrane Filters
Durable, reusable membranes withstand extreme chemical and thermal stress
  • 99.97% pure silver
  • Resistant to alcohols, fuels, hydrocarbons, oils, alkalis, ethers, and much more
  • Smooth surface—ideal for particle capture and easy identification
Use silver membrane filters in a variety of air and liquid filtration applications. Composed of pure metallic silver (99.97% pure silver), they make an ideal collection media for analysis of crystalline silica by X-ray diffraction and polyaromatic hydrocarbons (PAH). Silver membranes withstand a wide range of temperatures from cryogenic up to 1022°F (550°C). They can be autoclaved repeatedly and sterilized with steam or hot air for reuse.
122,000.00 - ₹128,000.00INR / Pkg of 50
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Description
Availability Pricing
COMPARE
25 Silver Filter, Silver, 0.45 µm, 25 mm; 50/PK
₹128,000.00INR / Pkg of 50
COMPARE
25 Silver Filter, Silver, 0.8 µm, 25 mm; 50/PK
₹122,000.00INR / Pkg of 50
COMPARE
37 Silver Filter, Silver, 0.8 µm, 37 mm; 25/PK
₹126,000.00INR / Pkg of 25