EBRO® TFX Compact Waterproof RTD Thermometer
The “must-have” premium handheld instrument 
Sometimes only the best is good enough. Tools used daily should be durable, reliable and best of quality. A pocket knife buff, for example, will use only an original Swiss Army Knife and never consider a less costly imitation. Why? Because there is no substitute for quality.
The same is true of the ebro TFX thermometer. The durable white ABS housing shows contamination at a glance and is designed to meet the demanding, robust needs of the food and catering industries. The entire instrument is watertight and meets all hygiene demands.
Whether used in food preparation or in a lab performing an experiment, cross contamination is a huge problem. Cross contamination can cause the material, even if it is initially safe, to be ruined, requiring extra time and money to resolve. Very often in daily practice to resolve contamination, the thermometer is cleaned at the sensor, however over time the instrument itself becomes a breeding ground for bacteria. Not so with the TFX, because not only can the probe be cleaned, the complete unit including connector between probe and instrument is watertight. You can even clean it under tap water!
TFX thermometers utilize high-precision RTD technology, reducing drifting, that is inherent with thermocouples, and the need for calibrations to an absolute minimum. The meter is powered by field replaceable lithium batteries, providing, even under low temperature conditions, an amazing battery lifetime of approx. 5 years. Providing an extremely accurate meter designed for applications requiring a high degree of reliability and repeatability.
These quality measuring instruments makes monitoring temperature in all phases of transport, storage and preparation a snap. Measurements are accomplished quickly and efficiently without interrupting normal work routines, and are ideal for use in supermarkets, restaurants, hospitals, laboratories and where food is processed.
The TFX is a premium thermometer tailored to the needs of food and laboratory professionals and offer important features:
- Easy to use
- Hygienic (dishwasher safe)
- Consistent, accurate results
- Reliable
- Calibration certificate