1. How often do you need to calibrate your ion-selective electrode (ISE)?
You should recalibrate at least once a week. You may need to recalibrate daily if you are in doubt of your results being accurate or reproducible.
2. After how long of being opened or made are standards good?
Stock standards will last at least six months before you need to discard them. Whereas diluted standards treated with ISA/pH buffer should be prepared weekly.
3. How do you store an ISE?
If you have a gas-sensing electrode, you should empty out the filling solution. If you have a liquid-sensing electrode, it is best to store it dry when it is not to be used within the next week or so. For those shorter periods of time, store it in a dilute standard approximating the sample concentration. You may need to add ISA/pH buffer when required.
4. Can your ISE system be temperature compensated?
Yes, it is possible, but somewhat difficult. First, you have to know the isopotential point for a given type of ISE system. Second, the concentration of the sample has to be similar in concentration to the isopotential point or else the temperature correction will be very inaccurate. Third, the temperature of the sample can not exceed the operational temperature range of the ISE. Fourth, very little isopotential point data for ISE's is available at this time. It is best to standardize and measure samples at the same temperature without using temperature compensation.
5. When measuring ions in-line continuously do you need to treat the sample?
Direct ion measurement is possible in many cases without ISA/pH buffer addition to the sample stream. However, fluoride, sulfide, ammonia, and sodium electrodes do require pH adjustment and must have ISA/pH buffer added to the sample stream.
6. Which standards should be used with an ISE?
The most obvious choice will be determined by what concentration units are desired. Also, if an electrode is being used to measure another ion, use a stock standard of the ion to be measured For an example, if you wanted to measure sulfate with a lead electrode, you would use a stock standard for sulfate.
7. Why buy a combination ISE instead of both a reference and a half cell ISE?
The advantages of a combination ISE are that you do not need to purchase an external reference electrode, it is more economically priced, and one less electrode to fit in your process.
Components of the Electrode System
Controllerpreform checkout procedure in instruction manual
Ion-Selective Electrodepreform electrode slope check, inspect for physical damage, polish, soak, or rebuild sensing membrane, monitor leak rate on reference side
Proper Calibration
Standardsprepare fresh standards by serial dilution
pH Adjustmentadjust pH to operational range of electrode
Sample Variable
Concentration Rangesee if sample is out of range
Pressurecheck if using wrong sample pressure
Temperatureverify sample temperature range
pHsee if correct sample pH
Interferencesare interferences present in the sample
Complexationare complexing agents in the sample