CavitationProcess in which small bubbles are formed and implode violently; occurs when NPSHa < NPSHr.
Dead HeadThe ability of a pump to continue running without damage when discharge is closed off. Only recommended for centrifugal pumps.
Density (specific weight of a fluid)Weight per unit volume, often expressed as pounds per cubic foot or grams per cubic centimeter.
Flooded SuctionLiquid flows to pump inlet from an elevated source by means of gravity. Recommended for centrifugal pump installations.
FlowA measure of the liquid volume capacity of a pump. Given in gallons per hour (GPH), gallons per minute (GPM), liters per minute (L/min), or milliliters per minute (mL/min).
FluidsInclude liquids, gases, and mixtures of liquids, solids, and gases. In this catalog, the terms fluid and liquid are both used to mean a pure liquid or a liquid mixed with gases or solids that acts essentially like a liquid in pumping applications.
HeadA measure of pressure, expressed in feet of head for centrifugal pumps. Indicates the height of a column of water being moved by the pump (without friction losses).
PressureThe force exerted on the walls of a tank, pipe, etc., by a liquid. Normally measured in pounds per square inch (psi).
PrimeCharge of liquid required to begin pumping action when liquid source is lower than pump. Held in pump by a foot valve on the intake line or by a valve or chamber within the pump.
SealsDevices mounted in the pump housing and/or on the pump shaft that prevent leakage of liquid from the pump.
Self-PrimingPumps that draw liquid up from below pump inlet (suction lift), as opposed to pumps requiring flooded suction.
Specific GravityThe ratio of the weight of a given volume of liquid to pure water. Pumping heavy liquids (specific gravity greater than 1.0) will require more drive horsepower.
Static Discharge HeadMaximum vertical distance (in feet) from pump to point of discharge with no flow.
StrainerA device installed in the inlet of a pump to prevent foreign particles from damaging the internal parts.
SumpA well or pit in which liquids collect below floor level; sometimes refers to an oil or water reservoir.
Total HeadSum of discharge head, suction lift, and friction loss.
Valves:
- Bypass ValveInternal to many pump heads that allow fluid to be recirculated if a given pressure limit is exceeded.
- Check ValveAllows liquid to flow in one direction only. Generally used in discharge line to prevent reverse flow.
- Foot ValveA type of check valve with a built-in strainer. Used at point of liquid intake to retain liquid in system, preventing loss of prime when liquid source is lower than pump.
- Relief ValveUsed at the discharge of a positive displacement pump. An adjustable, spring-loaded valve opens when a preset pressure is reached. Used to prevent excessive pressure buildup that could damage the pump or motor.
ViscosityThe "thickness" of a liquid or its ability to flow. Most liquids decrease in viscosity and flow more easily as they get warmer.