Solutions for your Vaccine Cold Chain
The vaccine cold chain is complicated with temperature challenges from end to end. Keep your vaccines safe, be compliant, and ensure your vaccine reaches the end user uncompromised when you use Traceable®, TraceableLIVE®, Cole-Parmer® and PolarSafe® products during storage and transport.
TEMPERATURE MONITORING
To manage an effective vaccine Cold Chain and be compliant, you must monitor the temperature while the vaccine is in storage and transit (and log the results for future review). This ensures your vaccine efficacy has not been compromised. Cloud-based technology allows you to manage this from a distance for added confidence.
Traceable® USB data logging thermometers for refrigerator/freezers, stores up to 10 unique alarm events; range –50 to 70°C
Traceable® USB data logging thermometer for ULT freezer uses a Pt RTD stainless steel probe; range –90 to 105°C
Connects to Wi-Fi and provides continuous data transmission to the TraceableLIVE® cloud-based interface for remote notifications; range –50 to 60°C
View current temperature conditions anywhere—cloud-based TraceableLIVE® interface sends email, text, and push notifications; range –50 to 60°C
Maintain compliance with easily generated vaccine reports using TraceableLIVE® cloud-based interface range –90 to 105°C
STORAGE
Although many vaccines at stored between +2° C and +8° C, there are also vaccines stored at -20° C and even newer vaccines that need to stored at ultra-low temperatures (-80° C). What is common in each situation is they have to reliably stay at the desired temperature and are organized for easy access and management — all the while ensuring the vaccine Cold Chain is not broken.
Built with temperature stability and energy savings in mind; Temperature range –86 to -40°C
Heated pressure equalization port for easy door opening; Temperature range –86 to -40°C
Adjustable full-extension sliding shelves provide efficient and customizable use of interior space and easy access to vaccines; range 2 to 10°C
Capacity 5.5 cu ft, exterior size is 26-7/8" wide, by 34-1/4" tall with a depth of 31-5/32"; range 2 to 10°C
Sealed to minimize moisture accumulation with sublids that provide added content protection; Temperature range -40°C to 0°C
TRANSPORTATION
In many applications, vaccine transportation will involve smaller amounts being moved short distances to secondary distributors, clinics, or simply to another building across the street because a refrigerator failed. This becomes a critical point to ensure the Cold Chain is not broken, and your vaccine is not compromised.
Durable polypropylene-insulated walls offer improved insulation for better vaccine protection during transporations
Insulated nylon bags provide an easy solution for transporting your samples
TraceableGO™ Data loggers record readings a minimum of every minute plus store up to 90 alarm events and can download data with free TraceableGO App
TraceableOne™ USB Temperature Data Loggers can be used for monitoring and recording temperatures during vaccine shipments