CPC (Colder)
CPC (Colder)
Colder (CPC) quick connect and disconnect solutions improve the functionality at your point of connection. Special features of CPC products include precise hose barbs for superior grip, built-in shutoff valves for preventing spills, and easy-to-use, push-button thumb latches for quick connections and disconnectionst. CPC couplings and fittings are perfect fo plastic tubing applications in the medical, biopharmaceutical, chemical handling, liquid cooling of electronics, industrial, printing, and food processing industries. Choose from standard product options and the ability to customize for your needs for non-valved, dripless, or sterile connections. When selecting fittings, make sure flow sizes of the bodies (female halves) and inserts (male halves) match.
APPLY FILTERS
filter byclear all
71 RESULTS
SORT BY
Showing 1 - 24 of 71
23>