CPC® (Colder) AseptiQuik® G Genderless Quick-Disconnect Fittings, Sanitary Adapter, High-Temperature Polycarbonate
Genderless sterile connectors simplify process integration while maximizing flexibility
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Robust construction provides reliable and repeatable fluid transfer
  • Sterilize by autoclave up to 130°C (266°F)
  • 0.5" nominal flow path size
The AseptiQuik® genderless connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, simply unsnap and flip down the protective tab cover, align connectors with the pull tabs hanging down, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.
All AseptiQuik connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot. High-temperature polycarbonate withstands autoclaving up to 130°C (266°F) for 60 minutes.
9,900.00 - ₹12,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Sterilization Method
Availability Pricing
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless, Sanitary, High-Temperature Polycarbonate, 3/4" Sanitary; 1/Ea 5011169 Mfr # AQG33012HT
COMPARE
Polycarbonate 3/4 Gamma irradiation up to 50 kGy or autoclave one cycle up to 130°C (266°F) for 60 minutes
₹9,900.00INR / Each
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless, Sanitary, High-Temperature Polycarbonate, 1-1/2" Sanitary; 1/Ea 5011171 Mfr # AQG33024HT
COMPARE
Polycarbonate 1-1/2 Gamma irradiation up to 50 kGy or autoclave one cycle up to 130°C (266°F) for 60 minutes
₹12,000.00INR / Each