Availability
VESPel
VESPel
DuPont™ Vespel/Graphite ferrules are crafted in a one-piece design that does not require a back ferrule. These reusable ferrules are recommended for GC/MS interface applications. We are a DuPont Vespel authorized distributor.