Vespel® Graphite Short Ferrules
Ideal for GC/MS interface applications
  • Manufactured of 60% Vespel and 40% Graphite
Vespel®/Graphite Ferrules are one-piece design requiring no back ferrule. Maximum temperature: 400°C. Recommended for GC/MS interface applications. Reusable – does not seize into column.
10,400.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
VESPel Graphite Short Ferrules Compatible with Agilent Inlets, 60% Vespel and 40% Graphite, 1/16" to 0.4 mm; 10/Pk 0199866 Mfr # 213164
COMPARE
Graphite Short Ferrules Compatible with Agilent Inlets, 1/16" to 0.4 mm
₹10,400.00INR / Pkg of 10
VESPel Graphite Short Ferrules Compatible with Agilent Inlets, 60% Vespel and 40% Graphite, 1/16" to 0.5 mm; 10/Pk 0199868 Mfr # 213165
COMPARE
Graphite Short Ferrules Compatible with Agilent Inlets, 1/16" to 0.5 mm
₹10,400.00INR / Pkg of 10
VESPel Graphite Short Ferrules Compatible with Agilent Inlets, 60% Vespel and 40% Graphite, 1/16" to 0.8 mm; 10/Pk 0199869 Mfr # 213168
COMPARE
Graphite Short Ferrules Compatible with Agilent Inlets, 1/16" to 0.8 mm
₹10,400.00INR / Pkg of 10