Cannon 9721-K68 Viscometer, universal size 300
Representative image only.
Cannon Mfr # 9721-K68 – Item # 9893436
Well suited for temperatures above 200°F
  • Calibrated in accordance with ASTM D446
  • 1 year warranty
PRODUCT OPTIONS
₹106,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.2%
  • Viscometer TypeCannon-Ubbelohde
  • Approximate Constant (cSt/sec)0.25
  • Sample Volume Needed (mL)11
  • Universal Size Number300
  • Min Viscosity Range (cSt)50
  • Max Viscosity Range (cSt)250
  • Warranty1 year
1 Year Warranty
Key Features
  • Calibrated in accordance with ASTM D446
  • 1 year warranty
More About this Item
Cannon-Ubbelohde viscometers are ideal for use in determining kinematic viscosities of transparent Newtonian liquids such as jet and hydraulic lubricants. Especially well suited for temperatures above 200°F. Viscometer constants do not vary with sample temperature. Viscometers are more durable than standard Ubbelohde viscometers. No kinetic energy corrections are required over a range of 0.5-100,000 cSt. Require an 11-mL sample volume.