Cannon-Ubbelohde Viscometers
Well suited for temperatures above 200°F
  • Calibrated in accordance with ASTM D446
Cannon-Ubbelohde viscometers are ideal for use in determining kinematic viscosities of transparent Newtonian liquids such as jet and hydraulic lubricants. Especially well suited for temperatures above 200°F. Viscometer constants do not vary with sample temperature. Viscometers are more durable than standard Ubbelohde viscometers. No kinetic energy corrections are required over a range of 0.5-100,000 cSt. Require an 11-mL sample volume.
106,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Approximate Constant (cSt/sec) Max Viscosity Range (cSt) Universal Size Number
Availability Pricing
Cannon 9721-K50 Viscometer, universal size 25 9893430 Mfr # 9721-K50
COMPARE
0.002 2 25
₹106,000.00INR / Each
Cannon 9721-K56 Viscometer, universal size 75 9893432 Mfr # 9721-K56
COMPARE
0.008 8 75
₹106,000.00INR / Each
Cannon 9721-K59 Viscometer, universal size 100 9893433 Mfr # 9721-K59
COMPARE
0.015 15 100
₹106,000.00INR / Each
Cannon 9721-K62 Viscometer, universal size 150 9893434 Mfr # 9721-K62
COMPARE
0.035 35 150
₹106,000.00INR / Each
Cannon 9721-K65 Viscometer, universal size 200 9893435 Mfr # 9721-K65
COMPARE
0.1 100 200
₹106,000.00INR / Each
Cannon 9721-K68 Viscometer, universal size 300 9893436 Mfr # 9721-K68
COMPARE
0.25 250 300
₹106,000.00INR / Each
Cannon 9721-K71 Viscometer, universal size 350 9893437 Mfr # 9721-K71
COMPARE
0.5 500 350
₹106,000.00INR / Each
Cannon 9721-K74 Viscometer, universal size 400 9893438 Mfr # 9721-K74
COMPARE
1.2 1200 400
₹106,000.00INR / Each
Cannon 9721-K77 Viscometer, universal size 450 9893439 Mfr # 9721-K77
COMPARE
2.5 2500 450
₹106,000.00INR / Each
Showing 1- 9 of 9