Cole-Parmer 384-Well Microplate Sealing Mat, Silicone, Square, Pre-Slit; 5/PK
Representative image only.
Cole-Parmer Item # 1292014
Ensure purity and prevent loss
  • Silicone reseals effectively to maintain sample purity
  • Individual well plugs eliminate cross-contamination
  • Pre-slit option available for easy pipetting
  • 384-well, square, pre-slit
PRODUCT OPTIONS
₹10,860.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells384
  • Well ShapeSquare, deep
  • MaterialSilicone
  • Description384-Well Microplate Sealing Mat, Silicone, Square, Pre-Slit; 5/PK
Key Features
  • Silicone reseals effectively to maintain sample purity
  • Individual well plugs eliminate cross-contamination
  • Pre-slit option available for easy pipetting
  • 384-well, square, pre-slit
More About this Item
Use these microplate sealing mats with well plates for storage prior to analysis. Silicone reseals after injections and pipetting for contamination-free sample preservation. Mats fit standard 96- and 384-well microplates.