Cole-Parmer® Microplate Sealing Mats
Ensure purity and prevent loss
  • Silicone reseals effectively to maintain sample purity
  • Individual well plugs eliminate cross-contamination
  • Pre-slit option available for easy pipetting
Use these microplate sealing mats with well plates for storage prior to analysis. Silicone reseals after injections and pipetting for contamination-free sample preservation. Mats fit standard 96- and 384-well microplates.
3,910.00 - ₹11,170.00INR / Pkg of 5
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Wells Well Shape Description
Availability Pricing
Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Round, Pre-Slit; 5/PK 1292006
₹5,710.00INR / Pkg of 5
COMPARE
96 Round, deep 96-Well Microplate Sealing Mat, Silicone, Round, Pre-Slit; 5/PK
Loading
₹5,710.00INR / Pkg of 5
Kinesis KX 96-Well Microplate Sealing Mat, Silicone, Low Profile Deep, Round, Pre-slit; 20/PK 1292007
Call For Price
COMPARE
96 Low-profile round 96-Well Microplate Sealing Mat, Silicone, Low Profile Deep, Round, Pre-slit; 20/PK
Loading
Call For Price
Cole-Parmer 384-Well Microplate Sealing Mat, Silicone, Square; 5/PK 1292012
₹6,330.00INR / Pkg of 5
COMPARE
384 Square, deep 384-Well Microplate Sealing Mat, Silicone, Square; 5/PK
Loading
₹6,330.00INR / Pkg of 5
Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Square, Pre-Slit; 5/PK 1292013
₹3,910.00INR / Pkg of 5
COMPARE
96 Square, deep 96-Well Microplate Sealing Mat, Silicone, Square, Pre-Slit; 5/PK
Loading
₹3,910.00INR / Pkg of 5
Cole-Parmer 384-Well Microplate Sealing Mat, Silicone, Square, Pre-Slit; 5/PK 1292014
₹10,860.00INR / Pkg of 5
COMPARE
384 Square, deep 384-Well Microplate Sealing Mat, Silicone, Square, Pre-Slit; 5/PK
Loading
₹10,860.00INR / Pkg of 5
Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Round; 5/PK 1292016
₹5,630.00INR / Pkg of 5
COMPARE
96 Round, deep 96-Well Microplate Sealing Mat, Silicone, Round; 5/PK
Loading
₹5,630.00INR / Pkg of 5
Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Round, Low Bleed; 5/PK 1292017
₹11,170.00INR / Pkg of 5
COMPARE
96 Round, deep 96-Well Microplate Sealing Mat, Silicone, Round, Low Bleed; 5/PK
Loading
₹11,170.00INR / Pkg of 5
Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Square; 5/PK 1292018
₹4,530.00INR / Pkg of 5
COMPARE
96 Square, deep 96-Well Microplate Sealing Mat, Silicone, Square; 5/PK
Loading
₹4,530.00INR / Pkg of 5
Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Shallow Well, Round, Pre-Slit; 5/PK 1292019
₹9,920.00INR / Pkg of 5
COMPARE
96 Round, shallow 96-Well Microplate Sealing Mat, Silicone, Shallow Well, Round, Pre-Slit; 5/PK
Loading
₹9,920.00INR / Pkg of 5
Showing 1- 9 of 9