Kinesis KX 96-Well Microplate Sealing Mat, Silicone, Low Profile Deep, Round, Pre-slit; 20/PK
Representative image only.
Cole-Parmer Item # 1292007
Ensure purity and prevent loss
  • Silicone reseals effectively to maintain sample purity
  • Individual well plugs eliminate cross-contamination
  • Pre-slit option available for easy pipetting
  • 96-well, low profile, round, pre-slit
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well ShapeLow-profile round
  • MaterialSilicone
  • Description96-Well Microplate Sealing Mat, Silicone, Low Profile Deep, Round, Pre-slit; 20/PK
Key Features
  • Silicone reseals effectively to maintain sample purity
  • Individual well plugs eliminate cross-contamination
  • Pre-slit option available for easy pipetting
  • 96-well, low profile, round, pre-slit
More About this Item
Use these microplate sealing mats with well plates for storage prior to analysis. Silicone reseals after injections and pipetting for contamination-free sample preservation. Mats fit standard 96- and 384-well microplates.