Cole-Parmer Blue-Sensitive Autoradiography Film, 5x7", 100/pk
Representative image only.
Cole-Parmer Item # 2170001
Get the results you need at an affordable price
  • Double emulsion film gives you sharp contrast and exemplary resolution for DNA sequencing
  • Single-sided film offers exceptional clarity and low background
PRODUCT OPTIONS
₹26,000.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Length (InchShort)5
  • Width (in)7
Key Features
  • Double emulsion film gives you sharp contrast and exemplary resolution for DNA sequencing
  • Single-sided film offers exceptional clarity and low background
More About this Item
Ideal for western blotting, DNA sequencing gels, chemiluminescent probes, gel shift analysis, and commonly used radioisotopes (32P, 125I). Use for manual or automatic development.