Cole-Parmer® Blue-Sensitive Autoradiography Film
Get the results you need at an affordable price
  • Double emulsion film gives you sharp contrast and exemplary resolution for DNA sequencing
  • Single-sided film offers exceptional clarity and low background
Ideal for western blotting, DNA sequencing gels, chemiluminescent probes, gel shift analysis, and commonly used radioisotopes (32P, 125I). Use for manual or automatic development.
26,000.00 - ₹33,100.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Blue-Sensitive Autoradiography Film, 5x7", 100/pk
₹26,000.00INR / Pkg of 100
COMPARE
Blue-Sensitive Autoradiography Film, 8 x 10", 100/pk
₹33,100.00INR / Pkg of 100