Cole-Parmer Cleanroom Polyester Swab, Double Layer Wipe Head, Notched, Light Green PP Handle; 100/PK
Representative image only.
Cole-Parmer Item # 3367604
Ideal for precision cleaning
  • Chemical resistance with a variety of solutions
  • Polypropylene handle ensures no additional contaminants
  • Suitable for ISO Class 5 cleanroom
  • Packed in large bag containing 2 bags of 50 swabs
PRODUCT OPTIONS
₹9,400.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyester
  • ColorLight Green
  • ComplianceISO Class 5 cleanroom
  • Swab Total Length (InchShort)1
  • Swab Total Length (cm)2.5
  • Length (in)5
  • Length (cm)12.75
  • DescriptionCleanroom Polyester Swab, Double Layer Wipe Head, Notched, Light Green PP Handle; 100/PK
  • WarrantyReplacement only
Key Features
  • Chemical resistance with a variety of solutions
  • Polypropylene handle ensures no additional contaminants
  • Suitable for ISO Class 5 cleanroom
  • Packed in large bag containing 2 bags of 50 swabs
More About this Item
These cleanroom swabs are ideal for precise cleaning, application of liquids or lubricants, and surface sampling validation. The swabs allow you to reduce the potential for lint and other residues in the cleanroom. The polypropylene handle helps to reduce any additional contaminants.
Applications include precision and medical device manufacturing.