Cole-Parmer® Cleanroom Swabs ISO Class 5
Ideal for precision cleaning
  • Chemical resistance with a variety of solutions
  • Polypropylene handle ensures no additional contaminants
  • Suitable for ISO Class 5 cleanroom
These cleanroom swabs are ideal for precise cleaning, application of liquids or lubricants, and surface sampling validation. The swabs allow you to reduce the potential for lint and other residues in the cleanroom. The polypropylene handle helps to reduce any additional contaminants.
7,300.00 - ₹16,200.00INR / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Swab Total Length (InchShort)
Availability Pricing
Cole-Parmer Cleanroom Polyester Swab, Double Layer Wipe Head, Notched, Light Green PP Handle; 100/PK 3367604
₹9,400.00INR / Pkg of 100
COMPARE
5 Polyester 1
Loading
₹9,400.00INR / Pkg of 100
Cole-Parmer Cleanroom Foam Swab, Open-Hole Head, Notched, Light Green PP Handle; 500/PK 3367605
₹16,200.00INR / Pkg of 500
COMPARE
3 21/64 Foam 43/64
Loading
₹16,200.00INR / Pkg of 500
Cole-Parmer Cleanroom Polyester Swab, Large Double Layer Wipe Head, Light Green PP Handle; 100/PK 3367611
₹7,300.00INR / Pkg of 100
COMPARE
5 5/32 Polyester 1
Loading
₹7,300.00INR / Pkg of 100