Cole-Parmer DCT-08 Manual Decapping Tool, 8 mm Dia
Representative image only.
Cole-Parmer Item # 9870684
Achieve a dependable and secure seal every time
  • Perfect for chromatography and general crimping and decapping applications
  • Remove 8 mm dia caps
PRODUCT OPTIONS
₹131,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cap Diameter (mm)8
  • DescriptionManual Decapping Tool, 8 mm Dia
Key Features
  • Perfect for chromatography and general crimping and decapping applications
  • Remove 8 mm dia caps
More About this Item
All crimpers and decappers are manufactured with a chemical-resistant finish and alloy crimping jaws. Crimpers provide a consistent seal for various cap sizes. Decapper offers a quick and easy seal removal.
The 11-, 13-, and 20-mm crimping tools feature a hex screw to adjust the crimping pressure and height, accommodating various neck designs and septa thicknesses.