Kinesis® Manual Crimpers and Decappers
Achieve a dependable and secure seal every time
  • Perfect for chromatography and general crimping and decapping applications
All crimpers and decappers are manufactured with a chemical-resistant finish and alloy crimping jaws. Crimpers provide a consistent seal for various cap sizes. Decapper offers a quick and easy seal removal.
The 11-, 13-, and 20-mm crimping tools feature a hex screw to adjust the crimping pressure and height, accommodating various neck designs and septa thicknesses.
101,000.00 - ₹490,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Cap Diameter (mm)
Availability Pricing
COMPARE
Manual Crimping Tool, 20 mm Dia 20
₹101,000.00INR / Each
COMPARE
Manual Decapping Tool, 20 mm Dia 20
₹107,000.00INR / Each
COMPARE
Manual Crimping Tool, 8 mm Dia 8
₹124,000.00INR / Each
COMPARE
Manual Decapping Tool, 8 mm Dia 8
₹131,000.00INR / Each
COMPARE
Manual Crimping Tool, 11 mm Dia 11
₹101,000.00INR / Each
COMPARE
Manual Decapping Tool, 11 mm Dia 11
₹109,000.00INR / Each
COMPARE
Manual Crimping Tool, 13 mm Dia 13
₹101,000.00INR / Each
COMPARE
Manual Decapping Tool, 13 mm Dia 13
₹107,000.00INR / Each
COMPARE
Manual Crimping Tool, 32 mm Dia 32
₹470,000.00INR / Each
COMPARE
Manual Decapping Tool, 32 mm Dia 32
₹490,000.00INR / Each
Showing 1- 10 of 10