Cole-Parmer DLF-400 UV Lamp, Timer, and Front Cover for 72" Vertical Flow Cabinet
Representative image only.
Cole-Parmer Item # 7890245
Accessories for Cole-Parmer® vertical laminar flow cabinets
  • Optimize laminar flow cabinet performance with UV lamp and base stand accessories
  • Suitable for use with Cole-Parmer® DLF-400 vertical laminar flow cabinets
  • Suitable for use with DLF-400 72" vertical flow cabinet
  • 1 year warranty
PRODUCT OPTIONS
₹116,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Hood Width (in)72
  • Warranty1 Year
1 Year Warranty
Key Features
  • Optimize laminar flow cabinet performance with UV lamp and base stand accessories
  • Suitable for use with Cole-Parmer® DLF-400 vertical laminar flow cabinets
  • Suitable for use with DLF-400 72" vertical flow cabinet
  • 1 year warranty
More About this Item
Choose from a wide range of accessories to enhance performance of your Cole-Parmer® DLF-400 vertical laminar flow cabinets. Accessories are available for use with the 48” or 72” cabinet, choose based on cabinet size you have.
The UV lamp is for decontamination of cabinet interior surfaces. This includes a timer, switch for UV lamp operation, and front cover. UV lamps must be purchased at the same time as the compatible cabinet since control panel with a UV key is factory installed in the cabinet.
The open frame base stand with casters in constructed of durable and corrosion resistant metal. This base provides a stable platform for cabinets and features locking casters for both stability and mobility when required.