Cole-Parmer® DLF-400 Vertical Laminar Flow Cabinet Accessries
Accessories for Cole-Parmer® vertical laminar flow cabinets
  • Optimize laminar flow cabinet performance with UV lamp and base stand accessories
  • Suitable for use with Cole-Parmer® DLF-400 vertical laminar flow cabinets
Choose from a wide range of accessories to enhance performance of your Cole-Parmer® DLF-400 vertical laminar flow cabinets. Accessories are available for use with the 48” or 72” cabinet, choose based on cabinet size you have.
The UV lamp is for decontamination of cabinet interior surfaces. This includes a timer, switch for UV lamp operation, and front cover. UV lamps must be purchased at the same time as the compatible cabinet since control panel with a UV key is factory installed in the cabinet.
The open frame base stand with casters in constructed of durable and corrosion resistant metal. This base provides a stable platform for cabinets and features locking casters for both stability and mobility when required.
114,000.00 - ₹262,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hood Width (in) Description
Availability Pricing
COMPARE
48
₹114,000.00INR / Each
COMPARE
72
₹116,000.00INR / Each
COMPARE
48
₹229,000.00INR / Each
COMPARE
72
₹262,000.00INR / Each