Cole-Parmer EX+ Adjustable Volume Pipettor, 0.1-2.0 uL; Each

Representative image only.
Cole-Parmer Item # 0791111
Chemical-resistant pipettor even handle solvents
  • Fully autoclavable
  • UV resistant for flexible sterilization
  • One-touch calibration
  • Enhanced ergonomics
  • 2 year warranty

PRODUCT OPTIONS

₹75,300.00INR / Each

₹17,600.00 - ₹25,900.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • SterileNo
  • Max Volume (µL)2
  • AutoclavableYes
  • Min Volume (µL)0.1
  • Min Volume (mL)0.0001
  • Max Volume (mL)0.002
  • Min Dispensing (mL)0.0001
  • Max Dispensing (mL)0.002
  • Increments (µL)0.01
  • Accuracy (%)±5.00% to ±3.00%
  • Number Of Channels1
  • Tip EjectorYes
  • Body MaterialStainless Steel (SS) and polypropylene (PP) plunger assembly
  • Warranty2 year
Two year warranty

Key Features

  • Fully autoclavable
  • UV resistant for flexible sterilization
  • One-touch calibration
  • Enhanced ergonomics
  • 2 year warranty

More About this Item

Chemical-resistant PTFE O-rings and seals allows these pipettors to be used with solvents. Fully autoclavable, adjustable volume pipettor features one-touch calibration and enhanced ergonomics. Enhanced UV resistance provides added protection from biohazardous elements. Easily adjust the pipette volume by turning the colored dial, and lock dial feature prevents accidental volume changes during use. Separate tip release button eliminates accidental release.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat