Cole-Parmer® EX-Plus Adjustable-Volume Pipettor
Chemical-resistant pipettor even handle solvents
  • Fully autoclavable
  • UV resistant for flexible sterilization
  • One-touch calibration
  • Enhanced ergonomics
Chemical-resistant PTFE O-rings and seals allows these pipettors to be used with solvents. Fully autoclavable, adjustable volume pipettor features one-touch calibration and enhanced ergonomics. Enhanced UV resistance provides added protection from biohazardous elements. Easily adjust the pipette volume by turning the colored dial, and lock dial feature prevents accidental volume changes during use. Separate tip release button eliminates accidental release.
75,300.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Max Volume (mL) Min Volume (mL)
Availability Pricing
COMPARE
Yes 0.002 0.0001
₹75,300.00INR / Each
COMPARE
Yes 0.01 0.0005
₹75,300.00INR / Each
COMPARE
Yes 0.02 0.002
₹75,300.00INR / Each
COMPARE
Yes 0.01 0.001
₹75,300.00INR / Each
COMPARE
Yes 0.2 0.02
₹75,300.00INR / Each
COMPARE
Yes 1 0.1
₹75,300.00INR / Each
COMPARE
Yes 5 1
₹75,300.00INR / Each
COMPARE
Yes 10 1
₹75,300.00INR / Each
Showing 1- 8 of 8