Cole-Parmer KX+ LCGC Total-Recovery Snap Top Vials, Glass, 11 mm Short Thread, 1.2 mL, PTFE/Silicone Septum; 100/pk
Representative image only.
Cole-Parmer Item # 9870168
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Total-recovery vials constructed of clear glass
  • PTFE/silicone septum
PRODUCT OPTIONS
₹24,800.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)1.2
  • Tube MaterialGlass
  • Tube ColorClear
  • Cap TypeSnap Top
  • Cap Diameter (mm)11
  • Septum TypePTFE/silicone
  • SterileNo
  • Special FeaturesTotal-Recovery
  • DescriptionLCGC Total-Recovery Snap Top Vials, Glass, 11 mm Short Thread, 1.2 mL, PTFE/Silicone Septum; 100/pk
Key Features
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Total-recovery vials constructed of clear glass
  • PTFE/silicone septum
More About this Item
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Snap-top vials come with snap caps that provide a secure seal to minimize evaporation, even with volatile samples. An audible click ensures a secure seal has been formed.
Glass vials are made of Type I, Class A 33-expansion borosilicate glass. Fused-insert vials are ideal for handling small-volume samples while delivering low residual volume (<8 µL). Total-recovery vials deliver maximum sample capacity (1.2 mL) with minimal residual volume (<1 µL)—use with bottom or side draw-port needles.