Cole-Parmer® KX+ LCGC Certified Snap Top Fused-Insert and Total Recovery Vials with Caps
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Snap-top vials come with snap caps that provide a secure seal to minimize evaporation, even with volatile samples. An audible click ensures a secure seal has been formed.
Glass vials are made of Type I, Class A 33-expansion borosilicate glass. Fused-insert vials are ideal for handling small-volume samples while delivering low residual volume (<8 µL). Total-recovery vials deliver maximum sample capacity (1.2 mL) with minimal residual volume (<1 µL)—use with bottom or side draw-port needles.
22,400.00 - ₹26,700.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Septum Type Special Features Volume (mL)
Availability Pricing
COMPARE
PTFE Fused Insert 0.3
₹22,400.00INR / Pkg of 100
COMPARE
PTFE Total-Recovery 1.2
₹22,900.00INR / Pkg of 100
COMPARE
PTFE/silicone Total-Recovery 1.2
₹24,800.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Total-Recovery 1.2
₹26,700.00INR / Pkg of 100