Cole-Parmer LiteSafe™ Light Blocking Centrifuge Tubes, Sterile, Polypropylene, Conical Bottom, 15 mL; 50/PK
Representative image only.
Cole-Parmer Item # 0634459
Designed to block 100% of UV rays for storage and centrifugation of light-sensitive samples
  • Completely opaque - protects light sensitive samples
  • Irradiation sterilized SAL 10-6
  • Safely centrifuge to 12,000 x g
  • DNase/RNase free, nonpyrogenic
PRODUCT OPTIONS
₹24,700.00INR / Pkg of 50
Often bought with...
Cole-Parmer® Disposable Plastic Serological Pipettes
9,200.00 - ₹24,000.00INR / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g8400;12000
  • SterileYes
  • AutoclavableYes
  • Tube Capacity (mL)15
  • Tube MaterialPolypropylene (PP)
  • Tube ColorBlack
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap StyleFlat Cap
  • Cap MaterialHigh-Density Polyethylene (HDPE)
  • GraduationsNone
  • Packaging50/Resealable Bag
  • DescriptionLight Blocking Centrifuge Tubes, Sterile, Polypropylene, Conical Bottom, 15 mL; 50/PK
Key Features
  • Completely opaque - protects light sensitive samples
  • Irradiation sterilized SAL 10-6
  • Safely centrifuge to 12,000 x g
  • DNase/RNase free, nonpyrogenic
More About this Item
For use with fluorescent probes, ethidium bromide, and other photosensitive samples, these conical bottom, polypropylene centrifuge tubes can block 100% of UV rays and are irradiation sterilized (SAL-10-6). They have a MAX RCF (maximum relative centrifugal force) of 12,000 x g and include flat HDPE caps. A frosted area for labeling is resistant to alcohol wiping.
Useful for the storage and centrifugation of light-sensitive samples, they can withstand temperatures from -80 °C (-112 °F) to 121 °C (250 °F). Packaged in resealable bags of 50 per bag, they are DNase/RNase-free, nonpyrogenic, and conform to USP Class VI standards.