Availability
Display Type
Max Power VDC (VDC)
Min Power VDC (VDC)
Panel Meters
The product searched is discontinued, please browse below for replacements.
Panel Meters
We offer both digital panel meters and analog panel meters. Digital panel meters provide efficiency and convenience for any lab or process. Analog meters are still popular for monitoring parameters that rapidly change. Many labs are finding the added functionality of digital panel meters of growing interest. Digital panel meters display temperature, pressure, voltage, current, and other signals from a wide array of sensors and transducers. Digital displays feature single- or dual-line readings depending on how many inputs the digital panel can accommodate. To control or send a signal to other process instruments, many panel meters feature output control signals and relays. Panel meters can be wall mounted or panel mounted. They are designed to save space without sacrificing performance. Many displays will change color to signal the process is out of the necessary range set during programming. Panel meters can be powered by wall power or by process system loop-power.