Cole-Parmer NIST-Traceable Viscosity Standard, S200; 500 mL
Representative image only.
Cole-Parmer Item # 9894480
Calibrated to a precision of ±0.2% of viscosity
  • Standards are NIST-traceable and certified to ASTM D2162 under ISO/IEC 17025 guidelines
PRODUCT OPTIONS
₹40,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Viscosity672 cSt at 68°F; 468 cSt at 77°F; 200 cSt (955 sus) at 100°F; 181 cSt at 104°F; 107 cSt at 122°F; 18 cSt (88.2 sus) at 210°F; 17 cSt at 212°F
Key Features
  • Standards are NIST-traceable and certified to ASTM D2162 under ISO/IEC 17025 guidelines
More About this Item
Calibrate kinematic and Saybolt viscosity equipment, both automatic and manual. A table on each bottle shows the approximate values for kinematic and Saybolt viscosity. Standards are supplied in 500-mL bottles and include full certification and a Material Data Safety Sheet.