Cole-Parmer® Viscosity Standards
Calibrated to a precision of ±0.2% of viscosity
  • Standards are NIST-traceable and certified to ASTM D2162 under ISO/IEC 17025 guidelines
Calibrate kinematic and Saybolt viscosity equipment, both automatic and manual. A table on each bottle shows the approximate values for kinematic and Saybolt viscosity. Standards are supplied in 500-mL bottles and include full certification and a Material Data Safety Sheet.
40,600.00 - ₹71,700.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Description Viscosity
Availability Pricing
COMPARE
NIST-Traceable Viscosity Standard, N.4; 500 mL 0.47 cSt at 68°F; 0.45 cSt at 77°F; 0.41 cSt at 100°F; 0.40 cSt at 104°F
Call For Price
COMPARE
NIST-Traceable Viscosity Standard, N.8; 500 mL 0.95 cSt at 68°F; 0.89 cSt at 77°F; 0.77 cSt at 100°F; 0.75 cSt at 104°F
Call For Price
COMPARE
NIST-Traceable Viscosity Standard, N1.0; 500 mL 1.3 cSt at 68°F; 1.2 cSt at 77°F; 1.0 cSt at 100°F; 0.97 cSt at 104°F
Call For Price
COMPARE
NIST-Traceable Viscosity Standard, S3; 500 mL 4.6 cSt at 68°F; 4.0 cSt at 77°F; 3.0 cSt at 100°F; 2.9 cSt at 104°F; 2.4 cSt at 122°F; 1.2 cSt at 210°F; 1.2 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, S6; 500 mL 10 cSt at 68°F; 8.7 cSt at 77°F; 6.0 cSt at 100°F; 5.7 cSt at 104°F; 4.5 cSt at 122°F; 1.9 cSt at 210°F; 1.9 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, N10; 500 mL 21 cSt at 68°F; 17 cSt at 77°F; 11 cSt at 100°F; 10 cSt at 104°F; 7.3 cSt at 122°F; 2.7 cSt at 210°F; 2.7 cSt at 212°F
₹71,700.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, S20; 500 mL 43 cSt at 68°F; 34 cSt at 77°F; 20 cSt (96.6 sus) at 100°F; 18 cSt at 104°F; 13 cSt at 122°F; 4.0 cSt at 210°F; 3.9 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, N35; 500 mL 77 cSt at 68°F; 59 cSt at 77°F; 35 cSt (152.1 sus) at 100°F; 29 cSt at 104°F; 20 cSt at 122°F; 5.3 cSt at 210°F; 5.2 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, S60; 500 mL 165 cSt at 68°F; 121 cSt at 77°F; 60 cSt (281 sus) at 100°F; 54 cSt at 104°F; 35 cSt at 122°F; 7.7 cSt at 210°F; 7.5 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, N100; 500 mL 372 cSt at 68°F; 268 cSt at 77°F; 128 cSt (592 sus) at 100°F; 114 cSt at 104°F; 70 cSt at 122°F; 13 cSt at 210°F; 13 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, S200; 500 mL 672 cSt at 68°F; 468 cSt at 77°F; 200 cSt (955 sus) at 100°F; 181 cSt at 104°F; 107 cSt at 122°F; 18 cSt (88.2 sus) at 210°F; 17 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, N350; 500 mL 1255 cSt at 68°F; 865 cSt at 77°F; 371 cSt at 100°F; 324 cSt at 104°F; 186 cSt at 122°F; 28 cSt (131.5 sus) at 210°F; 27 cSt at 212°F
₹47,800.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, S600; 500 mL 2184 cSt at 68°F; 1472 cSt at 77°F; 600 cSt at 100°F; 518 cSt at 104°F; 286 cSt (135.2 sfs) at 122°F; 37 cSt (174 sus) at 210°F; 37 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, N1000; 500 mL 4678 cSt at 68°F; 3089 cSt at 77°F; 1020 cSt at 104°F; 542 cSt at 122°F; 350 cSt at 140°F; 57 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, s2000; 500 mL 8323 cSt at 68°F; 5422 cSt at 77°F; 2000 cSt at 100°F; 1719 cSt at 104°F; 889 cSt at 122°F; 87 cSt (405 sus) at 210°F; 83.3 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, N4000; 500 mL 17,889 cSt at 68°F; 11,470 cSt at 77°F; 3448 cSt at 104°F; 1720 cSt at 122°F; 850 cSt at 140°F; 137 cSt at 210°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, S8000; 500 mL 34,931 cSt at 68°F; 22,383 cSt at 77°F; 8000 cSt at 100°F; 6710 cSt at 104°F; 3317 cSt at 122°F; 242 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, N15000; 500 mL 79,423 cSt at 68°F; 49,714 cSt at 77°F; 13,994 cSt at 104°F; 6650 cSt at 122°F; 3000 cSt at 140°F; 406 cSt at 212°F
₹40,600.00INR / Each
COMPARE
NIST-Traceable Viscosity Standard, S30000; 500 mL 84,687 cSt at 77°F; 28,079 cSt at 100°F; 23,570 cSt at 104°F; 11,058 cSt at 122°F; 628 cSt at 212°F
₹40,600.00INR / Each
Showing 1- 19 of 19