Cole-Parmer Spatula Macro Spoon, Stainless Steel,150mm; 3/PK
Representative image only.
Cole-Parmer Item # 0628705
Polished surface provides fast and easy cleaning
  • Chemical and corrosion resistant
  • Available in 6 sizes
PRODUCT OPTIONS
₹2,200.00INR / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)5 29/32
  • Width (in)1 1/10
  • Overall Length (cm)15
  • Width (cm)2.8
  • Material304 stainless steel
  • ColorMetalic
Key Features
  • Chemical and corrosion resistant
  • Available in 6 sizes
More About this Item
The AISI 304 grade stainless steel spatula/macro spoon combinations are chemical and corrosion resistant. One end features a macro spoon for medium and large size particles, while the other end offers a straight/flat spatula. Robust and seamless construction guarantees long life cycle.